SYNA791419
Synthware™ bushing adapter
female joint: ST/NS 14/20, male joint: ST/NS 19/22
Synonym(e):
Synthware A791419, glass bushing adapter
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Empfohlene Produkte
Verbindung
female joint: ST/NS14/20
male joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A791419
Allgemeine Beschreibung
Bushing adapters allow for compact assemblies of dissimilar joints.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)