Merck
SYNA691420
Synthware 90 degree vacuum adapter with glass stopcock
female joint: ST/NS 14/20
Synonym(e):
Synthware A691420, Flow control adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
female joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A691420
Verwandte Kategorien
Allgemeine Beschreibung
For regulating the input of gases or liquids into reaction systems. With 8 mm or 10 mm hose connection. With 8 mm or 10 mm hose connection.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
SYNA691420-1EA4061832189642