SYNA531420
Synthware™ inlet or thermometer adapter with hose connection
joint: ST/NS 14/20, O.D. 5.0-7.5 mm
Synonym(e):
Synthware A531420, thermometer inlet adapter
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A531420
AD
5.0-7.5 mm
Verwandte Kategorien
Allgemeine Beschreibung
Same as thermometer adapter but with 8mm or 10mm hose connection for N2/Ar introduction or vacuum. Supplied with one Nylon cap and one Viton® O-ring.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNA531420-1EA | 4061832189468 |