Merck
SYNA501922
Synthware gas inlet adapter
joint: ST/NS 19/22, O.D. × L 153 mm
Synonym(e):
Synthware A501922, gas inlet adapter, extended drip tube
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A501922
AD × L
153 mm
Verwandte Kategorien
Allgemeine Beschreibung
Inner joint with a sealed on lower tube extending 6” below the grind, having the same O.D. tubing on both ends of the grind.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)