Merck
SYNA501420
Synthware gas inlet adapter
joint: ST/NS 14/20, O.D. × L 8 mm × 153 mm
Synonym(e):
Synthware A501420, gas inlet adapter, extended drip tube
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A501420
AD × L
8 mm × 153 mm
Verwandte Kategorien
Allgemeine Beschreibung
Inner joint with a sealed on lower tube extending 6” below the grind, having the same O.D. tubing on both ends of the grind.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)