Merck
SYNA251914
Synthware enlarging adapter
top joint: ST/NS 19/22, bottom joint: ST/NS 14/20
Synonym(e):
Synthware A251914, glass connecting adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
bottom joint: ST/NS14/20
joint: ST/NS top19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A251914
Verwandte Kategorien
Allgemeine Beschreibung
Synthware enlarging adapter is used for connecting dissimilar standard taper joint sizes. The top joint (19/22) is larger that the bottom joint (14/20).
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)