Merck
DWKWCL1000AD-1
WHEATON® CELLine flasks for adherent cell culture
polystyrene, flask capacity (1,000 mL), case of 1 ea
Synonym(e):
Adherent Cell Culture Flasks, CELLine Flasks, Flasks for Adherent Cells
About This Item
UNSPSC-Code:
41122104
NACRES:
NB.13
Materialien
molded polypropylene neck
polystyrene
screw top plastic cap
Sterilität
sterile; γ-irradiated
Leistungsmerkmale
with cap
Verpackung
case of 1 ea
Hersteller/Markenname
WHEATON® WCL1000AD-1
Kolbenkapazität
(1,000 mL)
Allgemeine Beschreibung
CELLine AD Bioreactor Flask for high density adherent cell culture.
Media compartment with cell growth compartment for antibody and protein production.
Use with Hybridoma, Insect, CHO, NSO, HEK, BHK cell lines
  • Disposable and ready-to-use
  • High call density and high product concentration
  • Bulk media chamber allows for longer run times without media refreshing and reduces operation time
  • Decreases use of consumables
  • Cost-efficient, space saving, and stackable
  • No additional equipment required for operation
Leistungsmerkmale und Vorteile
  • Ideal for high density cell culture, designed to enhance small scale bio-production for antibody and protein generation
  • Available in 1000ml flask for suspension (CELLine 1000) or adherent (CELLine 1000-AD) cell culture and 350ml flask for suspension culture only
  • Disposable and ready-to-use
  • Multi-harvest operation ensures a cost efficient solution for antibody and protein production
  • Ideal for use with cell lines such as Hybridoma, CHO, HEK, BHK, Sf9
Rechtliche Hinweise
WHEATON is a registered trademark of Wheaton Industries, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)