CLS7715
Corning® thermometer stopper
rubber
Synonym(e):
Corning stopper
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.43
Empfohlene Produkte
Materialien
rubber
Hersteller/Markenname
Corning 7715
AD × ID
17 mm × 13 mm
Verwandte Kategorien
Allgemeine Beschreibung
This Corning rubber thermometer holder has a 17 mm O.D. and a 13 mm I.D. and is designed for use with the Corning CLS7711 mercury thermometer.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CLS7715-1EA | 4061825884448 |