CLS69471L
Pyrex® resin reaction kettles
capacity 1,000 mL
Synonym(e):
pyrex flask, pyrex with lid, pyrex with lids, Corning® 6947
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Empfohlene Produkte
Verpackung
case of 1 ea
Hersteller/Markenname
Corning 6947-1L
Verfügbarkeit
not available in EU
Kapazität
1,000 mL
Verwandte Kategorien
Allgemeine Beschreibung
Designed for viscous reaction materials. 500 and 1,000 mL kettles have interchangeable covers with four openings (24/40 joints). 2,000, 3,000 and 4,000 mL kettles have three openings with 24/40 joints and one with a 34/45 joint. Flanges of covers and bottoms are finely ground for a tight seal. Order clamp separately.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)