CLS401392
Corning® Modified Impeller Assembly (for use with probes) for Magnetically-Driven Proculture® Spinner Flask
fits flask, 8 L
Synonym(e):
equipment replacement accessory
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122104
NACRES:
NB.43
Empfohlene Produkte
Materialien
stainless steel shaft
Sterilität
non-sterile
Leistungsmerkmale
fits flask 8 L
Hersteller/Markenname
Corning 401392
Verwandte Kategorien
Allgemeine Beschreibung
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 8L Proculture Spinner Flasks. Narrower impeller paddle design allows use with probes to create a small bioreactor.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CLS401392-1EA | 4061825881102 |