CLS2400200
Pyrex® Liebig condenser with top and bottom joints
jacket L 200 mm, joint: ST/NS 19/38
Synonym(e):
Corning® 2400
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Empfohlene Produkte
Verbindung
joint: ST/NS19/38
Hersteller/Markenname
Corning 2400-200
Verfügbarkeit
not available in EU (Not available in EU countries)
Mantel L
200 mm
Allgemeine Beschreibung
With drip-tip. Large-capacity water jacket cools efficiently.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)