Merck
CLS1000PACK
PYREX® Griffin beakers
low form, assorted pack (contains 1 beaker of 5 different sizes)
Synonym(e):
Pyrex® beakers, Pyrex® glass beakers, beakers, glass beakers, Corning® 1000
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
Materialien
borosilicate glass
Leistungsmerkmale
graduations
Verpackung
case of 1
Hersteller/Markenname
Corning 1000-PACK
Verfügbarkeit
not available in EU
Allgemeine Beschreibung
Low Form (Corning 1000 series) Griffin beakers have the following features:
  • Graduated for approximate content
  • Double-graduated metric scale
  • Beakers have a spout
  • Extra-large area for marking
  • Uniform wall thickness
  • Heavy duty, industrial grade glass beakers
PYREX® assortment pack of Griffin Low form beakers, double scale, graduated A convenience pack of PYREX beakers contains one of each of the most popular sizes. Designed for the low volume user, a case contains one of each beaker: 50mL, 100mL, 250mL, 600mL and 1L. Packaged in a partitioned carton for safe transit and storage.
Sonstige Hinweise
Browse all glass beakers
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)