Merck
CLS430179
Corning® polypropylene beaker
sterile, size 250 mL , beaker, container and lid, case of 100
Synonym(e):
plastic beakers, polypropylene beakers, storage containers
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.13
Materialien
natural polypropylene (beaker)
orange high-density polyethylene (snap cap)
Sterilität
sterile
sterile
Leistungsmerkmale
container and lid
Verpackung
pack of 1
case of 100
Hersteller/Markenname
Corning 430179
Größe
250 mL , beaker
Allgemeine Beschreibung
Corning flexible polypropylene container with snap-on polyethylene lid serves as a beaker or storage container.
  • Container is graduated in both millilitres and ounces
  • Certified nonpyrogenic
Sonstige Hinweise
Browse all plastic beakers
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)