BR55618
BRAND® joint clip for dispensers with neck size 29/32
tilt measure stainless steel
Synonym(e):
Accessories, Compact Burettes, Dispensers, tilt measure
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Materialien
stainless steel
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 55618
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Joint Clip is specifically designed for dispensers with a neck size of 29/32, providing a secure and reliable connection for various laboratory applications. Made from high-quality stainless steel, this joint clip offers excellent durability and resistance to corrosion, ensuring long-lasting performance in demanding environments. Its precise design allows for easy attachment and detachment, facilitating efficient use in experiments and liquid handling. This essential accessory enhances the functionality of your dispensers, making it a valuable addition to any laboratory setup.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)