Merck
BR362038
BRAND® BLAUBRAND® ASTM 100 mL centrifuge tube
cylindrical, conical bottom, norm ASTM D91
Synonym(e):
ASTM, BLAUBRAND®, Centrifuge Tubes, Sample Storage, Tubes
About This Item
UNSPSC-Code:
41121703
NACRES:
NB.11
Materialien
borosilicate glass
conical bottom
cylindrical
Sterilität
non-sterile
Leistungsmerkmale
RCF 700 × g
Verpackung
pack of 2
Hersteller/Markenname
BRAND 362038
Länge
203 mm
Allgemeine Beschreibung
Withstands RCF up to 700. Design, accuracy, etc. to meet ASTM ("American Society for Testing and Materials") specifications.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)