Merck
BR27002
BRAND® SILBERBRAND ETERNA Graduated pipette, Class B, Type 2
1 mL with 0.01 mL graduations, not suitable for cotton plugs
Synonym(e):
ETERNA, Graduated Pipettes, Pipettes, SILBERBRAND
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Materialien
AR-Glass
Verpackung
pack of 12 ea
Hersteller/Markenname
BRAND 27002
Messbereich
accuracy: 0.01 mL
Kapazität
1.0 mL
Allgemeine Beschreibung
The BRAND® SILBERBRAND ETERNA graduated pipette is a Class B, Type 2 instrument made from AR-Glas® and complies with DIN EN ISO 835 standards. Designed for precision, it is calibrated to deliver (TD, Ex), ensuring accurate liquid transfer for various laboratory applications. This pipette combines durability with functionality, making it a reliable choice for professionals requiring high-quality measurement tools. Its clear graduation markings facilitate easy reading, enhancing user efficiency.
Rechtliche Hinweise
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)