Merck
BR134002
BRAND® non-return valve
for tube I.D. 6-9 mm
Synonym(e):
Non-return valve, Tubing
About This Item
UNSPSC-Code:
40141600
NACRES:
NB.35
Materialien
high-density polyethylene
Verpackung
pack of 10 ea
Hersteller/Markenname
BRAND 134002
Länge
80 mm
Verwandte Kategorien
Allgemeine Beschreibung
For tubing of inner- 6-9 mm. Length 80 mm. With FKM valve disc not suitable for pressure applications.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)