BAH367190000
SP Bel-Art® Quaverette Micro-Blade Vibrating Spatula
L 8.25 in. (21 cm), brown FEP blade
Synonym(e):
spoon
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.43
Empfohlene Produkte
Materialien
brown FEP blade
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art H36719-0000
Länge
8.25 in. (21 cm)
Allgemeine Beschreibung
Make accurate additions of precious materials as needed. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving you control over transfer from spatula to weighing dish.
Leistungsmerkmale und Vorteile
Available with a plain stainless steel blade or with a brown FEP coating
Coating complies with FDA regulations governing direct food contact applications; reduces transfer losses
Coating complies with FDA regulations governing direct food contact applications; reduces transfer losses
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)