BAH134160001
Dial calipers
L 222 mm
Alle Fotos(1)
About This Item
UNSPSC-Code:
41111700
NACRES:
NB.79
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art H13416-0001
Länge
222 mm
Allgemeine Beschreibung
Precision plastic calipers with Metric and English scales with 0.1 mm and 0.001 in. subdivisions. Use the thumb roller for precise measurements. Parallel jaws are 40 mm (1 ½ in.) deep.
- Non scratching, non-conductive and corrosion resistant
- Inside and outside jaws and round plastic rod depth gauge allow most measurements
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)