Principaux documents
EPM1282-9905
Platform for Eppendorf® New Brunswick Innova® 44/44R Incubator Shakers
for 125 mL Erlenmeyer flasks, dedicated, non-Interchangeable, aluminum
About This Item
Code UNSPSC :
41103816
Nomenclature NACRES :
NB.73
Matériaux
aluminum
Caractéristiques
dedicated
for 125 mL Erlenmeyer flasks
non-Interchangeable
Fabricant/nom de marque
Eppendorf® M1282-9905 (Global)
Capacité
(60 x 125 mL Erlenmeyer flasks)
Catégories apparentées
Description générale
Platforms to fit the Eppendorf New Brunswick 44 and 44 R incubated shakers.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG