Principaux documents
EPS44I200005
Eppendorf® Innova® S44i Shaker
incubated, AC/DC input 120 V
About This Item
Code UNSPSC :
41103811
Caractéristiques
incubated
Fabricant/nom de marque
Eppendorf® S44I200005
Description générale
The Innova S44i is a large capacity, temperature controlled (refrigerated) stackable shaker allowing the use of high-volume flasks in large numbers in a compact footprint. The iconic slide up door provides easy access to your samples. The Eppendorf X-Drive allows for shaking high loads at high speeds – even when stacked. When there is an instance of an unbalanced load, the X-Drive′s counterbalance adjustment allows for smooth shaking. The optional photosynthetic LED lightbank provides the ability to evenly light the entire platform across a wide intensity range – and it′s programmable. The VisioNize touch interface introduces onboard monitoring, user management and the ability to export data via USB; or network integration via ethernet. A comprehensive service offering including IQ/OQ is also available.
Product Information
The Innova S44i is a large capacity, temperature controlled (refrigerated) stackable shaker allowing the use of high-volume flasks in large numbers in a compact footprint. The iconic slide up door provides easy access to your samples. The Eppendorf X-Drive allows for shaking high loads at high speeds – even when stacked. When there is an instance of an unbalanced load, the X-Drive′s counterbalance adjustment allows for smooth shaking. The optional photosynthetic LED lightbank provides the ability to evenly light the entire platform across a wide intensity range – and it′s programmable. The VisioNize touch interface introduces onboard monitoring, user management and the ability to export data via USB; or network integration via ethernet. A comprehensive service offering including IQ/OQ is also available.
Maximum capacity
  • The Innova S44i has optimized chamber, platform, and features to maximize your options and capacity.
  • Up to 136 % more capacity than the previous Innova 44 with a smaller footprint
  • Up to 102 % higher flask capacity than competitor
  • Double- or triple-stack the Innova S44i to increase capacity without sacrificing more valuable lab space
Eppendorf X-Drive
  • The Eppendorf X-Drive is precisely engineered with five-eccentric shafts to balance even the heaviest load while providing consistent and smooth shaking.
  • The intelligent counterbalance technology of the Eppendorf X-Drive ensures vibration-free shaking even with uneven loads. This advanced technology senses imbalance and decelerates to a safe speed to protect you and your cultures.
Advanced control
The new touchscreen user interface of the Innova S44i offers more than just an easy operation. This multi-touch capacitive display makes all the information you need
easy to monitor and track.
  • Information is just one touch away
  • Intuitive operating, distinct design
  • Manage multiple users with different levels of authorization
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 