Principaux documents
EP022822228
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-35-6-30, (6 x 15 mL conical tube and 50 mL conical tubes )
About This Item
Code UNSPSC :
41103909
Caractéristiques
to hold 6 x 15 mL conical tubes
to hold 6 x 29.5x116-123 mm tubes
to hold 6 x 50 mL conical tubes
fixed angle rotor
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022822228 (NA)
Technique(s)
centrifugation: suitable
Capacité
(6 x 15 mL conical tube and 50 mL conical tubes )
Compatibilité
for use with centrifuge models Vacufuge Plus Concentrator
Description générale
Selection of rotors for the Eppendorf Vacufuge Plus.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG