Key Documents
Z532061
Sigma-Aldrich
Solid glass plug
O.D. 5/16 in.
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
D.E.
5/16 in.
7.9 mm
Description générale
Replace transfer tubing with this plug to make a leakproof Torion seal.
Informations légales
Torion is a trademark of Bibby Sterlin Ltd.
Pièce de rechange