Z341916
IKA® fixing clip attachment for HS/KS 260 basic and control shakers
AS 260.2 fixing clip attachment
Synonyme(s) :
flask clamps, holding clamps
About This Item
Code UNSPSC :
41103811
Nomenclature NACRES :
NB.73
Produits recommandés
Conditionnement
set of 1
Fabricant/nom de marque
IKA 3115500
Description générale
Note: This attachment (without fixing clips) is for shaking flasks, Erlenmeyer flasks and bottles with a round cross-section.
Informations légales
IKA is a registered trademark of IKA-Werke GmbH & Co. KG