Key Documents
WHA77700062
Whatman® UNIFILTER® plates
96 well, Lysate Clarification, well maximum volume 800 μL, lysate clarification media and 0.45?m polypropylene membrane
Synonyme(s) :
Z747041, micro plate, multiwell plate, well plates
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
Matériaux
polypropylene
polystyrene membrane (rigid high grade)
Stérilité
non-sterile
Caractéristiques
lysate clarification media and 0.45?m polypropylene membrane
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Whatman 7770-0062
Whatman Article No. 28420978 (US reference)
Volume maximum des puits
800 μL
Dimension de pores
0.45 μm
Type de liaison
non-treated surface
Description générale
  • Design prevents well to well crosstalk
  • Large variety of well volumes economizes on expensive consumables
  • Choice of filter media allows control of flow rates and retention characteristics
  • Standard overall dimensions fit most plate readers, auto samplers, robots
Application
The UNIFILTER products are convenient, ready to use, disposable filter bottom microplates. Available in 96 well formats, the UNIFILTER microplates offer a choice of filter media that work well with the most stringent applications. UNIFILTERS can be used to process 96 samples at a time. The unique drip director design allows for analysis of the filtrate or the retentate. UNIFILTERS are available in many different variations of well formats and range from 100 μL to 2 mL, using a wide variety of filtration media.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva