Principaux documents
CLS15002L
Corning® narrow mouth reagent bottle with Pyrex® ST stopper
capacity 2000 mL, stopper joint: 29
Synonyme(s) :
glass bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
clear bottle
cylindrical bottle
narrow-mouth bottle
Joint
joint: stopper29
Caractéristiques
cap
Fabricant/nom de marque
Corning 1500-2L
Capacité
2000 mL
Diam. × H
133 mm × 248 mm
Description générale
These Pyrex narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.