Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21099-U

Supelco

GC Stationary Phase

phase SE-30, bottle of 10 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
SB.54

Agency

meets requirements for USP G2

packaging

bottle of 10 g

parameter

temp. range at 50-300 °C

technique(s)

gas chromatography (GC): suitable

matrix active group

SE-30 phase

solubility

chloroform: soluble

column type

packed GC

General description

GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients. The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are a few parameters that should be considered before choosing a stationary phase. SE-30 is a dimethyl silicone elastomer. It is widely suitable as liquid phase got GLC.
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Application

SE-30 was used as stationary phase for the analysis of the thiohydantoins using GC. It may also be used as liquid phase in identification of the single polychlorinated biphenyls (PCB) components by comparing their retention indices, using high-resolution thin-film glass capillary gas chromatography coupled with electron-capture detector.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of polychlorinated biphenyls (PCB) by glass capillary gas chromatography.
Ballschmiter
Fresenius' Zeitschrift fur Analytische Chemie, 302, 20-31 (1980)
A C Moffat
Journal of chromatography, 113(1), 69-95 (1975-03-12)
The dimethyl silicone elastomer SE-30 has been chosen as the preferred liquid phase for the gas-liquid chromatographic analysis of drugs, and retention index data have been compiled for 480 drugs and commonly occurring chemicals such as plasticisers. The inter-laboratory variation
Analysis of amino acid phenylthiohydantoins by gas chromatography.
J J Pisano et al.
The Journal of biological chemistry, 244(20), 5597-5607 (1969-10-25)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service