Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4667

Sigma-Aldrich

GDF-5 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

CDMP-1, Cartilage-Derived Morphogenetic Protein-1, Cartilage-derived morphogenetic protein-1, LAP4, Radotermin, SYNS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~26.8 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GDF5(8200)

General description

GDF-5 acts as an inhibitor of nerve tissue growth. It is produced as a homodimer, non-glycosylated polypeptide chain containing 2 X 117 amino acids with a molecular weight of 26.8 kDa. To enable bacterial expression of rhGDF-5 we removed the N-terminal sequence Ala-Pro- Leu- Thr and added the Lys.

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in 20 Mm HOAc to a concentration of 0.1 to 1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunfeng Zhao et al.
Clinical orthopaedics and related research, 472(9), 2569-2578 (2014-06-08)
Adhesions and poor healing are complications of flexor tendon repair. The purpose of this study was to investigate a tissue engineering approach to improve functional outcomes after flexor tendon repair in a canine model. Flexor digitorum profundus tendons were lacerated
Zhou Pei et al.
Archives of biochemistry and biophysics, 560, 27-35 (2014-08-01)
Adipocyte differentiation is key to determining the number of adipocytes during the development of obesity. Recent studies have shown that growth differentiation factor-5 (GDF5) promotes brown adipogenesis, however its role in white adipogenesis is still uncertain. The aim of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service