Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3022

Sigma-Aldrich

CTGFL-WISP-2 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

CCN5, CT-58, Connective Tissue Growth Factor-Like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

10-20 ng/mL

mol wt

24.3 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... WISP2(8839)

General description

CTGFL/WISP-2 is a 28.6 kDa protein that belongs to the CCN family of cysteine rich regulatory proteins. Members of this family stimulate mitosis, adhesion, apoptosis, extracellular matrix production, growth arrest, and migration of multiple cell types. The protein is expressed in primary osteoblasts, fibroblasts, ovary, testes, and heart. In addition to promoting adhesion of osteoblasts, CTGFL/WISP-2 inhibits osteocalcin production, as well as binding of fibrinogen to integrin receptors. Recombinant human CTGFL/WISP-2 is a 24.8 kDa protein of 228 amino acid residues.

Biochem/physiol Actions

CTGFL/WISP-2 is a 28.6 kDa protein that belongs to the CCN family of cysteine rich regulatory proteins. Recombinant human CTGFL/WISP-2 is a 24.8 kDa protein of 228 amino acid residues.

Sequence

MQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAF

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Note: Do not raise to neutral pH. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kumar et al.
The Journal of biological chemistry, 274(24), 17123-17131 (1999-06-08)
We have identified and cloned a novel connective tissue growth factor-like (CTGF-L) cDNA from primary human osteoblast cells encoding a 250-amino acid single chain polypeptide. Murine CTGF-L cDNA, encoding a polypeptide of 251 amino acids, was obtained from a murine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service