Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2031

Sigma-Aldrich

CTF1 human

recombinant, expressed in insect cells, ≥70% (SDS-PAGE)

Synonym(s):

CTF, CTF/NFI, NF-1, NF-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

biological source

human

recombinant

expressed in insect cells

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~56.4 kDa

packaging

pkg of 5 μg

concentration

500 μg/mL

color

colorless to clear

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CTF-1(1489)

Biochem/physiol Actions

The CCAAT-box binding protein 1 (CTF1, also called NF-I or NF-1) is a proline-rich transcription activator. It selectively activates transcription of genes that contain GCCAAT consensus sequence in promoter region, including human Ha-Ras, alpha-globin, hsp70 and many other cellular and viral genes. Cloning and functional characterization of the protein revealed that CTF1 contains an N-terminal DNA binding domain and a C-terminal proline-rich activation domain. CTF1 was shown to interact with the general transcription factor IIB (TFIIB) and facilitate the recruitment of TFIIB to TBP-DNA complexes, with the forkhead thyroid transcription factor TTF2 to regulate hormone–dependent transcription of thyroperoxidase gene, as well as other receptor proteins to synergize hormone receptor-responsive activation.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Mermod et al.
Cell, 58(4), 741-753 (1989-08-25)
Human CTF/NF-I consists of a family of CCAAT box binding proteins that activate both transcription and DNA replication. Analysis of cDNA mutants expressed in E. coli and Drosophila cells reveals that the N-terminal portion of CTF-1 is sufficient for site-specific
K A Jones et al.
Cell, 48(1), 79-89 (1987-01-16)
Transcription factor CTF, which is responsible for selective recognition of eukaryotic promoters that contain the sequence CCAAT, was purified to apparent homogeneity by sequence-specific DNA affinity chromatography. Binding sites for CTF in the human Ha-ras and alpha-globin promoters were highly
C Santoro et al.
Nature, 334(6179), 218-224 (1988-07-21)
The CTF/NF-I group of cellular DNA binding proteins recognizes the sequence GCCAAT and is implicated in eukaryotic transcription as well as DNA replication. Molecular analysis of human CTF/NF-I complementary DNA clones reveals multiple messenger RNA species containing alternative coding regions
Alejandro Aranda et al.
Cellular and molecular life sciences : CMLS, 71(23), 4637-4651 (2014-05-06)
We report a new method to generate high-expressing mammalian cell lines in a quick and efficient way. For that purpose, we developed a master cell line (MCL) containing an inducible alphavirus vector expressing GFP integrated into the genome. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service