MilliporeSigma
  • A family of human CCAAT-box-binding proteins active in transcription and DNA replication: cloning and expression of multiple cDNAs.
A family of human CCAAT-box-binding proteins active in transcription and DNA replication: cloning and expression of multiple cDNAs.
Nature (1988-07-21)
C Santoro, N Mermod, P C Andrews, R Tjian
ABSTRACT
The CTF/NF-I group of cellular DNA binding proteins recognizes the sequence GCCAAT and is implicated in eukaryotic transcription as well as DNA replication. Molecular analysis of human CTF/NF-I complementary DNA clones reveals multiple messenger RNA species containing alternative coding regions, apparently as a result of differential splicing. Expression and functional analysis establish that individual gene products can bind to GCCAAT recognition sites and serve both as promoter-selective transcriptional activators and as initiation factors for DNA replication.
Sigma-Aldrich
CTF1, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)