Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0222

Sigma-Aldrich

PTPmu Active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

PTPRL1, PTPRM, Protein tyrosine phosphatase mu, R-PTP-MU, hR-PTPu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

56.2 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPRM(5797)

General description

Protein tyrosine phosphatase. Catalytic domain, (amino acids 839-1100), (Gene bank accession number BC051651) with N-terminal GST tag, MW = 56.2 kDa, expressed in an E. coli expression system.

Application

Useful for the study of enzyme kinetics, regulation, and to dephosphorylate target substrates.

Unit Definition

One unit will hydrolyze 1 nmol p-nitrophenyl phosphate per minute at pH 7.4 and 30°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 75 mM NaCl, 0.05% Tween-20, 50% glycerol, 2 mM EDTA, 1 mM DTT, and 10 mM glutathione.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service