Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4702060

Monoclonal Anti-CD10 PE-Cyanine7 produced in mouse

new

Synonym(s):

Atriopeptidase, CALLA, Enkephalinase, MME, Neprilysin, Neutral endopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

PE-Cy7 conjugate

Quality Level

antibody product type

primary antibodies

clone

MEM-78, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MME(4311)

General description

CD10 (neutral endopeptidase – NEP, common acute lymphocytic leukemia antigen – CALLA, membrane metallo-endopeptidase – MME, enkefalinase) is a 100-kDa cell surface zinc metalloprotease, cleaving peptide bonds on the N-terminus of hydrophobic amino acids and inactivating multiple physiologically active peptids. CD10 is expressed on various normal cell types, including lymphoid precursor cells, germinal center B lymhocytes, and some epithelial cells, and its expression level serves as a marker for diagnostics of many carcinomas. CD10 is also a differentiation antigen for early B-lymphoid progenitors in the B-cell differentiation pathway and has a key role in regulation of growth, differentiation and signal transduction of many cellular systems.

Specificity

The antibody MEM-78 reacts with an extracellular epitope CD10 antigen (CALLA - Common acute lymphatic leukemia antigen), a 100 kDa type II integral membrane protein.

Immunogen

NALM-6 human pre-B cell line

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with activated tandem dye of R-phycoerythrin-cyanine 7 (PE-Cy7) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service