Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501496

Sigma-Aldrich

Anti-NDUFS2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Complex 1, mitochondrial respiratory chain, 49-KD subunit, NADH dehydrogenase (ubiquinone) Fe-S protein 2, NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase), NADH-ubiquinone oxidoreductase NDUFS2 subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, mouse, bovine, human, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDUFS2(4720)

General description

NADH: ubiquinone oxidoreductase core subunit S2 (NDUFS2) is a 49kDa protein. The gene encoding it is localized on human chromosome 1q23.

Immunogen

Peptide with sequence SRGVRQWQPDVE, from the internal region of the protein sequence according to NP_004541.1; NP_001159631.1

Biochem/physiol Actions

NADH: ubiquinone oxidoreductase core subunit S2 (NDUFS2) is a subunit of complex I, which is involved in the oxidation of NADH and electron transfer to ubiquinone.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mapping to 1q23 of the human gene (NDUFS2) encoding the 49-kDa subunit of the mitochondrial respiratory Complex I and immunodetection of the mature protein in mitochondria.
Procaccio V
Mammalian Genome, 9(6), 482-484 (1998)
Human Ind1, an Iron-Sulfur Cluster Assembly Factor for Respiratory Complex I?
Alex D
Molecular and Cellular Biology, 29 (2009)
Understanding mitochondrial complex I assembly in health and disease?
Masakazu Mimaki
Biochim. Biophys. Acta Gen. Subj. (2012)

Global Trade Item Number

SKUGTIN
SAB2501496-100UG4061831451184

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service