Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB2108447

Sigma-Aldrich

Anti-BAX

IgG fraction of antiserum

Synonym(s):

Anti-CLECSF10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

21 kDa

species reactivity

bovine, human, dog, pig

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

accession no.

NM_138761

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

B-cell lymphoma 2 associated X (BAX), also known as CLECSF10 (C-type lectin superfamily member 10), is located on human chromosome 19q13. BAX is a proapoptotic protein and is a member of Bcl-2 protein family.

Immunogen

Synthetic peptide directed towards the N terminal region of human BAX

Application

Anti-BAX has been used in Western blotting.

Biochem/physiol Actions

B-cell lymphoma 2 associated X (BAX) promotes cell death and plays a vital role in regulation of apoptosis. BAX gene may act as a negative regulator of autophagy in CRC (colorectal cancer) development.

Sequence

Synthetic peptide located within the following region: MDGSGEQPRGGGPTSSEQIMKTGALLLQGFIQDRAGRMGGEAPELALDPV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced radiation effect on SMCC7721 cells through endoplasmic reticulum stress induced by C225-GNPs in vitro and in vivo
Zhu C, et al.
Oncology Letters, 15, 4221-4228 (2018)
Cytoprotective Peptide Humanin Binds and Inhibits Proapoptotic Bcl-2/Bax Family Protein BimEL
Luciano F, et al.
The Journal of Biological Chemistry, 280, 15825-15835 (2005)
Immediate early up-regulation of bax expression by p53 but not TGF beta 1: a paradigm for distinct apoptotic pathways.
Selvakumaran M, et al.
Oncogene, 9, 1791-1798 (1994)
The BAX gene as a candidate for negative autophagy-related genes regulator on mRNA levels in colorectal cancer
Gil J, et al.
Medical Oncology (Northwood, London, England), 34 (2017)
The substitution of Proline 168 favors Bax oligomerization and stimulates its interaction with LUVs and mitochondria.
Simonyan L, et al.
Biochimica et Biophysica Acta, 1859, 1144-1155 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service