Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1410711

Sigma-Aldrich

Anti-IL9 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HP40, IL-9, P40

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 15.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL9(3578)

General description

The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness. (provided by RefSeq)

Immunogen

IL9 (NP_000581.1, 1 a.a. ~ 144 a.a) full-length human protein.

Sequence
MLLAMVLTSALLLCSVAGQGCPTLAGILDINFLINKMQEDPASKCHCSANVTSCLCLGIPSDNCTRPCFSERLSQMTNTTMQTRYPLIFSRVKKSVEVLKNNKCPYFSCEQPCNQTTAGNALTFLKSLLEIFQKEKMRGMRGKI

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Sun et al.
Hypertension in pregnancy, 39(3), 228-235 (2020-04-25)
Our study aimed to investigate IL9 and IL9R expression in preeclampsia and assess their effects on trophoblast biological behaviors. IL9 and IL9R expression of placenta tissue were evaluated by immunochemistry and q-PCR. Using transwell, CCK-8, and tubule formation assays measured
Rafael Ribeiro Almeida et al.
British journal of cancer, 123(4), 534-541 (2020-06-06)
Host-microbiota interactions shape T-cell differentiation and promote tumour immunity. Although IL-9-producing T cells have been described as potent antitumour effectors, their role in microbiota-mediated tumour control remains unclear. We analysed the impact of the intestinal microbiota on the differentiation of
Atsuo Nakamura et al.
Archives of biochemistry and biophysics, 693, 108587-108587 (2020-09-19)
Polyamines produced by both prokaryotes and eukaryotes are bioactive substances with pleiotropic effects. Accumulating evidence has demonstrated that polyamines contribute to anti-inflammatory responses by suppressing the expression of proinflammatory cytokines in mononuclear cells and macrophages. However, the effects of polyamines
Wanzhou Zhao et al.
Journal of immunology research, 2020, 4184380-4184380 (2020-04-08)
T helper (Th) cells orchestrate allergic lung inflammation in asthma pathogenesis. Th9 is a novel Th cell subset that mainly produces IL-9, a potent proinflammatory cytokine in asthma. A 7-amino acid peptide (7P) of the hypervariable region 1 (HVR1) of
Juan Xiao et al.
Frontiers in pharmacology, 11, 859-859 (2020-06-27)
The incidence of pancreatitis (AP) is increasing and there is no specific treatment available. Intracellular digestive enzyme activation is a key event in the pathogenesis of AP downstream of cytosolic calcium overload and impaired autophagy. Siraitia grosvenorii (Swingle) was used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service