MilliporeSigma
  • Decreased placental IL9 and IL9R in preeclampsia impair trophoblast cell proliferation, invasion, and angiogenesis.
Decreased placental IL9 and IL9R in preeclampsia impair trophoblast cell proliferation, invasion, and angiogenesis.
Hypertension in pregnancy (2020-04-25)
Yi Sun, Siyu Liu, Rong Hu, Qiongjie Zhou, Xiaotian Li
ABSTRACT
Our study aimed to investigate IL9 and IL9R expression in preeclampsia and assess their effects on trophoblast biological behaviors. IL9 and IL9R expression of placenta tissue were evaluated by immunochemistry and q-PCR. Using transwell, CCK-8, and tubule formation assays measured invasion, proliferation and angiogenesis of trophoblast with adding IL9 or anti-IL9R antibody. IL9 and IL9R levels were significantly decreased in preeclampsia. IL9 improved trophoblast activities. Blocking IL9/IL9R resulted in decreased proliferation, invasion, and tube-formation capability of trophoblast. IL9 and IL9R contribute to the pathogenesis of preeclampsia. IL9/IL9R signaling provides a new potential therapeutic target for preventing preeclampsia.