Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB1403990

Sigma-Aldrich

Monoclonal Anti-IRF4, (C-terminal) antibody produced in mouse

clone 2D1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

LSIRF, MUM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IRF4(3662)

General description

Interferon regulatory factor 4 (IRF4), also known as multiple myeloma oncogene-1 (MUM1), is a transcription factor. It is encoded by the gene mapped to human chromosome 6p25.3. The protein is specifically expressed in lymphocytes.

Immunogen

IRF4 (NP_002451, 342 a.a. ~ 451 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LCNDRPNKLERDQTCKLFDTQQFLSELQAFAHHGRSLPRFQVTLCFGEEFPDPQRQRKLITAHVEPLLARQLYYFAQQNSGHFLRGYDLPEHISNPEDYHRSIRHSSIQE

Biochem/physiol Actions

Interferon regulatory factor 4 (IRF4) plays a vital role in regulation of immunoglobulin class switch recombination and plasma cell differentiation. In addition, it is also implicated in maintaining homeostasis of both mature B and mature T lymphocytes. Mutation in the gene is associated with the risk of developing chronic lymphocytic leukemia (CLL).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prognostic significance of interferon regulating factor 4 (IRF4) in node-negative breast cancer.
Heimes AS, et al.
Journal of Cancer Research and Clinical Oncology, 143(7), 1123-1131 (2017)
Specificity of IRF4 Translocations for Primary Cutaneous Anaplastic Large Cell Lymphoma: a Multicenter Study of 204 Skin Biopsies
Wada DA, et al.
Modern Pathology, 24(4), 596-596 (2011)
Verification that common variation at 2q37.1, 6p25.3, 11q24.1, 15q23, and 19q13.32 influences chronic lymphocytic leukaemia risk
Crowther?Swanepoel D, et al.
British Journal of Haematology, 150(4), 473-479 (2010)
H W Mittrücker et al.
Science (New York, N.Y.), 275(5299), 540-543 (1997-01-24)
Lymphocyte-specific interferon regulatory factor (LSIRF) (now called IRF4) is a transcription factor expressed only in lymphocytes. Mice deficient in IRF4 showed normal distribution of B and T lymphocyes at 4 to 5 weeks of age but developed progressive generalized lymphadenopathy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service