Recommended Products
form
powder
color
white
solubility
DMSO: soluble 1 mg/mL
antibiotic activity spectrum
Gram-negative bacteria
Gram-positive bacteria
Mode of action
enzyme | inhibits
storage temp.
−20°C
InChI
1S/C7H10N2O2S.C2H4O2/c8-5-6-1-3-7(4-2-6)12(9,10)11;1-2(3)4/h1-4H,5,8H2,(H2,9,10,11);1H3,(H,3,4)
InChI key
UILOTUUZKGTYFQ-UHFFFAOYSA-N
Biochem/physiol Actions
Mafenide is a bacteriostatic antibiotic with a broad activity against Gram positive, Gram negative bacteria and Clostridia. Mafenide acetate penetrates eschar and prevents heavy colonization in burn wounds.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Annals of burns and fire disasters, 24(3), 132-137 (2012-03-08)
Histological and clinical studies of wound healing were performed in comparable cases of fresh partial-thickness burns treated with honey dressing or mafenide acetate in two groups of 50 randomly allocated patients. Of the patients with honey-treated wounds, 84% showed satisfactory
Archivos argentinos de pediatria, 110(4), 298-303 (2012-08-04)
Topical agents are widely used in the care of burn patients; however the efficacy to prevent local infections and/or sepsis has not been clearly established in studies with a high level of evidence. This systematic review was conducted to assess
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service