Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I7408

Sigma-Aldrich

Anti-phospho-Integrin β3 (pTyr773) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
mouse, rat, human
citations:
1
technique(s):
western blot: 0.1-1.0 μg/mL using K562 cells

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 130 kDa

species reactivity

mouse, rat, human

technique(s)

western blot: 0.1-1.0 μg/mL using K562 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

target post-translational modification

phosphorylation (pTyr773)

Gene Information

human ... ITGB3(3690)
mouse ... Itgb3(16416)
rat ... Itgb3(29302)

General description

The gene ITGB3 (integrin subunit β 3) encodes the β subunit of integrins, which are transmembrane α/β heterodimers. There are 18 α subunits and 8 β subunits identified in mammals that associate with each other to form 24 different heterodimers. Based on the type of β3 subunit, the heterodimers can be grouped into subfamilies.

Immunogen

synthetic phosphopeptide derived from the region of human integrin β3 receptor that contains tyrosine 773. The sequence is conserved in human, mouse, and rat.

Biochem/physiol Actions

Integrins function in various cellular processes, such as tissue migration during embryogenesis, cellular adhesion, cancer metastases, and lymphocyte helper and killer cell functions. The β3 subunit is expressed along with αv and αIIb subunits in platelets to form the subfamilies αvβ3 and αIIbβ3. The receptor αvβ3 facilitates platelet adhesion to osteopontin and vitronectin in vitro. But, it is yet to be determined if it functions in platelet adhesion in vivo. The αvβ3 receptor functions as a receptor for fibrinogen, vWF, fibronectin, and vitronectin and participates in platelet aggregation. Any defects in the number or function of this receptor may result in the inhibition of platelet aggregation, leading to the bleeding disorder Glanzmann thrombasthenia.

Physical form

Solution in Dulbecco′s PBS, containing 1 mg/mL BSA, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel S Bennett
The Journal of clinical investigation, 115(12), 3363-3369 (2005-12-03)
The platelet integrin alpha(IIb)beta(3) is required for platelet aggregation. Like other integrins, alpha(IIb)beta(3) resides on cell surfaces in an equilibrium between inactive and active conformations. Recent experiments suggest that the shift between these conformations involves a global reorganization of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service