Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H4040

Sigma-Aldrich

HydroMatrix Peptide Cell Culture Scaffold

24 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

synthetic

Quality Level

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

HydroMatrix has been shown to support improved growth and migration in the proliferation of many cell types, including neural stem cells, neurons, glia, astrocytes, fibroblasts, and keratinocytes.

Components

HydroMatrix is a peptide nanofiber 3-dimensional scaffold that promotes cell growth and migration. It utilizes specific peptides that self-assemble from fluid precursors into highly crosslinked peptide hydrogels when presented with increased temperatures or ionic strengths. By inducing a rapid transformation from solution to gel, a peptide nanofiber scaffold is generated. Adjusting the concentration of the HydroMatrix solution, researchers can control the flexibility of the 3-D architecture to meet their application′s specific needs.

Caution

The lyophilized product at -20°C and is stable for many years at this temperature. The hydrogel should be stored at -70°C to maximize its active span. The 1% (w/v) stock solution should be stored -20°C and is stable for 3-6 months.

Preparation Note

This product is supplied as a lyophilized powder. A 1% (w/v) stock solution at 10 mg/mL can be prepared with addition of sterile water, resulting in a rigid gel. A 0.5% (w/v) solution will lead to a softer gel that is more suitable for cell migration and angiogenesis. Lower concentrations in the range of 0.15%-0.25% are more suitable for many cell types including neurons. In aqueous solution, HydroMatrix has a pH of 2.5 and should not be added directly to cells without prior addition of a buffer or medium. Once the hydrogel has formed, do not attempt to mix the gel any further, as this risks destroying the hydrogel integrity. Manipulation of the cells should be carefully performed to avoid disruption of the scaffold.

Legal Information

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service