Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G7035

Sigma-Aldrich

Anti-Rat IgG (whole molecule)–Gold antibody produced in goat

affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

gold conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

aqueous glycerol suspension

particle size

10 nm (colloidal gold)

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Rat IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rat IgG conjugated to a detectable substrate is a useful tool for the analysis of target proteins. Anti-Rat IgG (whole molecule)-Gold antibody is specific for rat immunoglobulins and does not react with human serum proteins.

Immunogen

Rat IgG purified from normal rat serum

Application

Anti-Rat IgG (whole molecule)-Gold antibody is suitable for use in dot blot assay.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in Tris buffered saline, pH 8.2, containing 1% (w/v) bovine serum albumin, 30% (v/v) glycerol, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan Van Blerkom et al.
Reproductive biomedicine online, 33(4), 458-475 (2016-08-16)
The oolemma of the second metaphase mouse oocyte has a relatively large domain in the peri-polar body region that is non-permissive for sperm attachment. In this study, certain biochemical components of this non-permissive domain were examined and compared with permissive
Mathieu Bangert et al.
The Journal of infectious diseases, 205(9), 1399-1407 (2012-03-30)
New treatments against severe bacterial infections are needed because the response to antibiotic treatment is slow in acute settings and is becoming less effective owing to the emergence of antibiotic-resistant pathogens. P4-mediated antibody therapy offers a unique treatment strategy that
Patricia M Tilney et al.
PloS one, 9(5), e96405-e96405 (2014-05-09)
Secondary pollen presentation is a well-known phenomenon in the Rubiaceae with particularly conspicuous pollen presenters occurring in the tribe Vanguerieae. These knob-like structures are formed by a modification of the upper portion of the style and stigma, together known as
Chao Huang et al.
Diabetes, 67(8), 1639-1649 (2018-06-06)
Diabetic retinopathy (DR) is a microvascular complication of diabetes and is the leading cause of vision loss in working-age adults. Recent studies have implicated the complement system as a player in the development of vascular damage and progression of DR.
Agata Leszczuk et al.
Scientific reports, 10(1), 20621-20621 (2020-11-28)
Arabinogalactan proteins (AGPs) are ubiquitous cell wall and plasma membrane components and are characterised by extensive glycosylation and heterogeneity of their carbohydrate and protein units. The aim of the study was to evaluate the structural features of AGPs present in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service