Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU019961

Sigma-Aldrich

MISSION® esiRNA

targeting human TNFAIP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGCGGAAAGCTGTGAAGATACGGGAGAGAACTCCAGAAGACATTTTTAAACCTACTAATGGGATCATTCATCATTTTAAAACCATGCACCGATACACACTGGAAATGTTCAGAACTTGCCAGTTTTGTCCTCAGTTTCGGGAGATCATCCACAAAGCCCTCATCGACAGAAACATCCAGGCCACCCTGGAAAGCCAGAAGAAACTCAACTGGTGTCGAGAAGTCCGGAAGCTTGTGGCGCTGAAAACGAACGGTGACGGCAATTGCCTCATGCATGCCACTTCTCAGTACATGTGGGGCGTTCAGGACACAGACTTGGTACTGAGGAAGGCGCTGTTCAGCACGCTCAAGGAAACAGACACACGCAACTTTAAATTCCGCTGGCAACTGGAGTCTCTCAAATCTCAGGAATTTGTTGAAACGGGGCTTTGCTATGATACTCGGAACTGGAATGATGAATGGGACAATCTTATCAAAATGGCTTCCACAGACACACCCATG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhipeng Li et al.
Brain, behavior, and immunity, 79, 228-235 (2019-02-11)
Neuroinflammation is now recognized to be a feature of many neurological disorders. More accumulated evidences suggested chrysin which was contained in honey, propolis, vegetables, fruits and plants can exert biological activities including anti-neuroinflammatory effects. However, the precise molecular mechanisms of
Hongjun Zhao et al.
Rheumatology (Oxford, England), 56(5), 835-843 (2017-02-06)
TNF-α-induced protein 3 ( TNFAIP3 ) is one of the major SLE susceptibility genes involved in the regulation of inflammatory responses through modulation of the nuclear factor-κB (NF-κB) pathway. We aim to assess TNFAIP3 expression in CD4 + T cells
Wenjing Feng et al.
Biochemical and biophysical research communications, 482(4), 1107-1113 (2016-12-05)
The innate immune response provides the first line of defense against viruses and other pathogens by responding to specific microbial molecules. A20 is a cytoplasmic ubiquitin-editing protein that negatively regulates the retinoic acid-inducible gene I (RIG-I)-mediated activation of interferon regulatory
Meng Qin et al.
Frontiers in pharmacology, 8, 953-953 (2018-01-10)
Atherosclerosis (AS) is a chronic inflammatory disease and endothelial cell injury is the initial event. In this study, we investigated the protective effects of ginsenoside F1 (GF1) on AS and the potential molecular mechanisms of ox-LDL induced endothelial injury. ApoE-/-
Ming-Yang Li et al.
Oncotarget, 7(48), 79914-79924 (2016-11-09)
The regulatory B cells (Breg) are important in the body immunity. The differentiation process of Breg is not fully understood yet. Ubiquitin A20 has immune regulatory functions. This study aims to investigate the role of A20 in the regulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service