Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

45338

Supelco

Benodanil

PESTANAL®, analytical standard

Synonym(s):

2-Iodobenzanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10INO
CAS Number:
Molecular Weight:
323.13
Beilstein:
2725018
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Ic1ccccc1C(=O)Nc2ccccc2

InChI

1S/C13H10INO/c14-12-9-5-4-8-11(12)13(16)15-10-6-2-1-3-7-10/h1-9H,(H,15,16)

InChI key

LJOZMWRYMKECFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A C Udeogalanya
Beitrage zur tropischen Landwirtschaft und Veterinarmedizin, 20(4), 467-474 (1982-01-01)
When applied at the juvenile growth stage, benodanil significantly reduced the occurrence of rust (Puccinia bordei) on barley plants. This was found in the susceptible varieties Midas, Zephyr, and Mazurka as well as in the slow-rusting varieties Tyra, Emir, and
A C Udeogalanya
Beitrage zur tropischen Landwirtschaft und Veterinarmedizin, 22(1), 91-94 (1984-01-01)
Leaf washing after application of benodanil and oxycarboxin reduced the protectant effect more than the eradicant effect. Oxycarboxin penetrated more rapidly than did benodanil . Residues of benodanil (WP) on the leaf surfaces can be absorbed again later in the
Xiao-Lei Zhu et al.
ChemMedChem, 9(7), 1512-1521 (2014-03-29)
Succinate-ubiquinone oxidoreductase (SQR, EC 1.3.5.1), also known as mitochondrial respiratory complex II or succinate dehydrogenase (SDH), catalyzes the oxidation of succinate to fumarate as part of the tricarboxylic acid cycle. SQR has been identified as a novel target of a large family

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service