Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

31580

Supelco

Terbufos-sulfone

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21O4PS3
CAS Number:
Molecular Weight:
320.43
Beilstein:
2279368
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)SCS(=O)(=O)C(C)(C)C

InChI

1S/C9H21O4PS3/c1-6-12-14(15,13-7-2)16-8-17(10,11)9(3,4)5/h6-8H2,1-5H3

InChI key

DWZSTEUTHNUVQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terbufos-sulfone is one of the major metabolite of organophosphorus pesticide terbufos, found to be toxic for living beings even at very low levels.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Terbufos-sulfone may be used as a pesticide reference standard for the determination of the analyte:
  • In fruits, vegetables and milk samples by gas chromatography (GC) coupled with ion trap mass spectrometry (MS/MS).
  • In water samples by a dispersive solid-phase extraction method based on the use of multiwalled carbon nanotubes.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonel Córdoba Gamboa et al.
Environmental research, 184, 109243-109243 (2020-02-23)
Pesticides used in agriculture may expose populations living nearby. Costa Rica is a major banana-exporting country, its production depends on extensive pesticide use. To evaluate environmental pesticide exposure, we measured levels of current-use pesticides in air and dust from 12
Catherine B Choung et al.
Environmental pollution (Barking, Essex : 1987), 159(10), 2648-2655 (2011-07-09)
To examine the link between corn agriculture and the observed decline of the endangered southern bell frog (SBF), the effects of two corn crop pesticides on larval growth and development were investigated. Tadpoles were exposed to terbufos sulfone (10 μg/L)
Analysis of pesticides residues in environmental water samples using multiwalled carbon nanotubes dispersive solid-phase extraction.
Gonzalez?Curbelo MA
Journal of Separation Science, 36(3), 556- 563 (2013)
Analysis of pesticide residues in fruits, vegetables, and milk by gas chromatography/tandem mass spectrometry.
Sheridan RS and Meola JR
Journal of AOAC (Association of Official Analytical Chemists) International, 82(3), 982- 990 (1999)
B T Bowman et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 17(5), 447-462 (1982-01-01)
Terbufos, t.sulfoxide and t.sulfone (5 micrograms ml-1) were incubated in natural, sterilized natural and distilled water, with initial pH values of 8.8, 8.8 and 6.0, respectively, at 20 degrees C. First-order disappearance was observed for the three compounds. Rates in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service