Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB1879

Sigma-Aldrich

Anti-Caspase 8 Antibody

Chemicon®, from rabbit

Synonym(s):

MACH1, FLICE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

monkey, bovine, rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP8(841)
mouse ... Casp8(12370)

General description

Caspase-8 is a 55 kDa cytosolic protein that is synthesized as an inactive proenzyme. Activation of caspase-8 involves a two-step proteolysis: the cleavage of caspase-8 to generate a 43 kDa fragment and a 12 kDa which is further processed to 10 kDa, and then the p43 is then cleaved to yield p26 and the release of the active site containing p18.

Specificity

AB1879 detects 55 and 43 kDa proteins, corresponding to the proform and large subunit of caspase-8. The antibody also detects human recombinant caspase-8 protein on SDS-PAGE immunoblots.

Immunogen

recombinant, full length, purified human caspase 8

Application

Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Caspases

Enzymes & Biochemistry
This Anti-Caspase 8 Antibody is validated for use in WB for the detection of Caspase 8.
Western blot: 0.5 - 4 μg/mL

Optimal working dilutions must be determined by end user.

Target description

55 and 43 kDa

Linkage

Replaces: 04-573; 04-574

Physical form

ImmunoAffinity Purified
Liquid in PBS, pH 7.2, containing 30% glycerol, 0.5% BSA and 0.01% thiomerosal. Affinity Purified immunoglobulin.

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Skin/Tumor, Embryo

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxia-inducible factor-2? regulates Fas-mediated chondrocyte apoptosis during osteoarthritic cartilage destruction.
Ryu, JH; Shin, Y; Huh, YH; Yang, S; Chun, CH; Chun, JS
Cell Death and Differentiation null
Margarita Ramirez-Ortega et al.
European journal of pharmacology, 566(1-3), 34-42 (2007-05-01)
Our aim in performing this study was to analyze in vivo the cell death mechanism induced by toxic doses of digitalis compounds on guinea-pig cardiomyocytes. We analyzed three study groups of five male guinea pigs each. Guinea pigs were intoxicated
Lyssavirus matrix protein induces apoptosis by a TRAIL-dependent mechanism involving caspase-8 activation.
Kassis, R; Larrous, F; Estaquier, J; Bourhy, H
Journal of virology null
Proliferation and apoptosis in mammary epithelium during the rat oestrous cycle.
R Strange, K C Westerlind, A Ziemiecki, A-C Andres, R Strange, K C Westerlind et al.
Acta Physiologica (Oxford, England) null
Characterization of events associated with apoptosis/anoikis induced by snake venom metalloproteinase BaP1 on human endothelial cells.
Cecilia Diaz, Lorena Valverde, Oscar Brenes, Alexandra Rucavado, Jose Maria Gutierrez
Journal of Cellular Biochemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service