Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21119

Sigma-Aldrich

Thiosemicarbazide

for synthesis

Synonym(s):

Thiosemicarbazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH5N3S
CAS Number:
Molecular Weight:
91.14
MDL number:
UNSPSC Code:
12352111
EC Index Number:
201-184-7

Quality Level

form

solid

potency

9.16 mg/kg LD50, oral (Rat)
2200 mg/kg LD50, skin (Rabbit)

pH

5-7 (20 °C, 50 g/L in H2O)

mp

178-180 °C (decomposition)

bulk density

500‑600 kg/m3

storage temp.

15-25°C

InChI

1S/CH5N3S/c2-1(5)4-3/h3H2,(H3,2,4,5)

InChI key

BRWIZMBXBAOCCF-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 3

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service