Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CF0027

Sigma-Aldrich

Phenylsulfanyltetrafluoropropionic acid Cs salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H5F4O2SCs
MDL number:
UNSPSC Code:
12352101

form

solid

reaction suitability

reaction type: C-C Bond Formation

Application

Phenylsulfanyltetrafluoropropionic acid Cs salt is as a less hygroscopic, crystalline source of phenylsulfanyltetrafluoropropionic acid. A phenylsulfanyltetrafluoroethyl moiety incorporated in the substrate can be treated with tributyltin hydride and generate the corresponding fluoroalkyl radical. If the substrate lacks any olefins, it can be reduced to tetrafluoroethyl group whereas if the substrate contains an olefin in the correct spatial orientation, it can lead to an intramolecular cyclization affording tetrafluorinated cyclic structures.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The fluoroalkylation toolbox now includes Togni reagents, hypervalent iodine perfluoroalkylation reagents, fluoroalkyl bromides, silanes, carboxylates, and sulfonyl fluorides for late stage fluoroalkylation.

The fluoroalkylation toolbox now includes Togni reagents, hypervalent iodine perfluoroalkylation reagents, fluoroalkyl bromides, silanes, carboxylates, and sulfonyl fluorides for late stage fluoroalkylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service