Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808059

Sigma-Aldrich

Copper

nanowires, diam. × L ~100 nm (±20 nm) × 10~20 μm, ~5 mg/mL in hexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
Molecular Weight:
63.55
UNSPSC Code:
12352200
NACRES:
NA.23

form

nanowires

concentration

~5 mg/mL in hexane

diam. × L

~100 nm (±20 nm) × 10~20 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Copper nanowires find applications in flexible electronics, transparent electrodes, photocatalysis, solar cells and as shape memory material [1, 2, 3].

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-14.8 °F

Flash Point(C)

-26 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service