Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

692921

Sigma-Aldrich

Bis[di-(tert-butyl)(4-trifluoromethylphenyl)phosphine]palladium(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H44Cl2F6P2Pd
CAS Number:
Molecular Weight:
757.93
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

230 °C (D)

functional group

phosphine

SMILES string

Cl[Pd]Cl.CC(C)(C)P(c1ccc(cc1)C(F)(F)F)C(C)(C)C.CC(C)(C)P(c2ccc(cc2)C(F)(F)F)C(C)(C)C

InChI

1S/2C15H22F3P.2ClH.Pd/c2*1-13(2,3)19(14(4,5)6)12-9-7-11(8-10-12)15(16,17)18;;;/h2*7-10H,1-6H3;2*1H;/q;;;;+2/p-2

InChI key

ATVRGWRBZLLSJD-UHFFFAOYSA-L

Application

Catalyst for Suzuki Coupling reaction of heteroaryl chlorides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anil S Guram et al.
The Journal of organic chemistry, 72(14), 5104-5112 (2007-06-07)
The new air-stable PdCl2[PR2(Ph-R')]2 complexes, readily prepared from commercial reagents, exhibit unique efficiency as catalysts for the Suzuki-Miyaura coupling reactions of a variety of heteroatom-substituted heteroaryl chlorides with a diverse range of aryl/heteroaryl boronic acids. The coupling reactions catalyzed by
Anil S Guram et al.
Organic letters, 8(9), 1787-1789 (2006-04-21)
[reaction: see text] New air-stable PdCl(2){P(t)Bu(2)(p-R-Ph)}(2) (R = H, NMe(2), CF(3),) complexes represent simple, general, and efficient catalysts for the Suzuki-Miyaura cross-coupling reactions of aryl halides including five-membered heteroaryl halides and heteroatom-substituted six-membered heteroaryl chlorides with a diverse range of

Articles

A-Phos Palladium Complexes for Efficient Suzuki Coupling

A-Phos Palladium Complexes for Efficient Suzuki Coupling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service